accessibility menu, dialog, popup

UserName

CAS RN 69335-91-7

IUPAC Name: 2-(4-{[5-(trifluoromethyl)pyridin-2-yl]oxy}phenoxy)propanoic acid
Synonyms: fluazifop
Molecular Weight (g/mol): 327.26
Molecular Formula: C15H12F3NO4
InChi Key: YUVKUEAFAVKILW-UHFFFAOYNA-N
SMILES: CC(OC1=CC=C(OC2=CC=C(C=N2)C(F)(F)F)C=C1)C(O)=O
Molecular Weight (g/mol) 
  • (1)
  • (2)
Quantity 
  • (1)
  • (1)
Solvent or Matrix 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results