accessibility menu, dialog, popup

UserName

ONOOO

CAS RN 698-63-5

IUPAC Name: 5-nitrofuran-2-carbaldehyde
Synonyms: 5-nitro-2-furaldehyde
Molecular Weight (g/mol): 141.08
Molecular Formula: C5H3NO4
InChi Key: SXINBFXPADXIEY-UHFFFAOYSA-N
SMILES: [O-][N+](=O)C1=CC=C(O1)C=O
Molecular Weight (g/mol) 
  • (5)
  • (3)
Quantity 
  • (2)
  • (1)
  • (1)
  • (2)
  • (2)
Percent Purity 
  • (1)
  • (2)
  • (3)
Physical Form 
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (3)
  • (1)
  • (5)
  • (3)
  • (2)
  • (1)
  • (1)
  • (2)
  • (2)
  • (1)
  • (2)
  • (3)
  • (3)
  • (3)