missing translation for 'onlineSavingsMsg'
Learn More

5-Nitro-2-furaldehyde, TRC

Product Code. 30468191
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30468191 5 g
30466425 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30468191 Supplier Toronto Research Chemicals Supplier No. TRCN4959305G
 Restricted Item

This item requires a price update prior to being ordered.

Request a Price for this item here.

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 698-63-5
Molecular Formula C5 H3 N O4
Molecular Weight (g/mol) 141.08
Synonym 5-Nitro-2-furfural
IUPAC Name 5-nitrofuran-2-carbaldehyde
SMILES [O-][N+](=O)c1oc(C=O)cc1

Specifications

Quantity 5 g
InChI Formula InChI=1S/C5H3NO4/c7-3-4-1-2-5(10-4)6(8)9/h1-3H
Formula Weight 141.0062
Chemical Name or Material 5-Nitro-2-furaldehyde
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.