missing translation for 'onlineSavingsMsg'
Learn More

5-Nitro-2-furaldehyde, 97%

Product Code. 10752601 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
10 g
50 g
Packaging:
Amber glass bottle
Glass bottle
Unit Size:
10g
1g
50g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10752601 10 g Glass bottle 10g
10518663 1 g Amber glass bottle 1g
10696924 50 g Glass bottle 50g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10752601 Supplier Thermo Scientific Acros Supplier No. 173860100

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 698-63-5 | C5H3NO4 | 141.08 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 698-63-5
MDL Number MFCD00003230
InChI Key SXINBFXPADXIEY-UHFFFAOYSA-N
Synonym 5-nitro-2-furaldehyde, 5-nitrofurfural, nitrofurfural, 2-furancarboxaldehyde, 5-nitro, 5-nitrofurfuraldehyde, 5-nitro-2-furfural, 2-furaldehyde, 5-nitro, 5-nitrofuraldehyde, 5-nitro-2-furancarboxaldehyde, furfural, 5-nitro
PubChem CID 12774
ChEBI CHEBI:34459
IUPAC Name 5-nitrofuran-2-carbaldehyde
SMILES C1=C(OC(=C1)[N+](=O)[O-])C=O

Specifications

CAS Min % 96.0
CAS Max % 100.0
Melting Point 34.0°C to 35.0°C
Color Yellow to Brown
Density 1.3400g/mL
Flash Point 33°C
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Packaging Glass bottle
Molecular Formula C5H3NO4
Refractive Index 1.5905
Quantity 10 g
Beilstein 17,IV,4459
Specific Gravity 1.34
Solubility Information Solubility in water: slightly soluble
Molecular Weight (g/mol) 141.08
Formula Weight 141.08
Percent Purity 97%
Physical Form Crystalline Low Melting Solid
Chemical Name or Material 5-Nitro-2-furaldehyde
Show More Show Less
compliance-icons
Product Identifier
  • 5-Nitro-2-furaldehyde
Signal Word
  • Warning
Hazard Category
  • Flammable solid Category 2
Hazard Statement
  • H228-Flammable solid.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P403+P233-Store in a well-ventilated place. Keep container tightly closed.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.