accessibility menu, dialog, popup

UserName

CAS RN 7005-72-3

IUPAC Name: 1-chloro-4-phenoxybenzene
Synonyms: 4-chlorophenyl phenyl ether
Molecular Weight (g/mol): 204.65
Molecular Formula: C12H9ClO
InChi Key: PGPNJCAMHOJTEF-UHFFFAOYSA-N
SMILES: ClC1=CC=C(OC2=CC=CC=C2)C=C1
Quantity 
  • (1)
  • (1)
  • (1)
Concentration 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results