accessibility menu, dialog, popup

UserName

CAS RN 702662-50-8

ONSOHNOClF

CAS RN 702662-50-8

IUPAC Name: 4-[(2-chloro-6-fluorophenyl)methyl]-N-[(furan-2-yl)methyl]-3-oxo-3,4-dihydro-2H-1,4-benzothiazine-6-carboxamide
Synonyms: 4-[(2-chloro-6-fluorophenyl)methyl]-N-(furan-2-ylmethyl)-3-oxo-2H-1,4-benzothiazine-6-carboxamide
Molecular Weight (g/mol): 430.88
Molecular Formula: C21H16ClFN2O3S
InChi Key: NAGKYJATVFXZKN-UHFFFAOYSA-N
SMILES: FC1=C(CN2C(=O)CSC3=C2C=C(C=C3)C(=O)NCC2=CC=CO2)C(Cl)=CC=C1
Color 
  • (4)
Grade 
  • (4)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results