accessibility menu, dialog, popup

UserName

CAS RN 71566-07-9

CH3NONBr

CAS RN 71566-07-9

IUPAC Name: 5-(4-bromophenyl)-3-methyl-1,2,4-oxadiazole
Synonyms: 5-(4-bromophenyl)-3-methyl-1,2,4-oxadiazole
Molecular Weight (g/mol): 239.07
Molecular Formula: C9H7BrN2O
InChi Key: AVXXUFMZRHQRTK-UHFFFAOYSA-N
SMILES: CC1=NOC(=N1)C1=CC=C(Br)C=C1
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results