missing translation for 'onlineSavingsMsg'
Learn More

5-(4-bromophenyl)-3-methyl-1,2,4-oxadiazole, 97%, Thermo Scientific™

Product Code. 11330687
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
Packaging:
Amber Glass Bottle
Unit Size:
1g
250mg
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
11330687 250 mg Amber Glass Bottle 250mg
10717865 1 g Amber Glass Bottle 1g
10386224 5 g Amber Glass Bottle 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11330687 Supplier Thermo Scientific Maybridge Supplier No. CC50910CB
 Restricted Item
Estimated Shipment: 01-10-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Chemical Identifiers

CAS 71566-07-9
Molecular Formula C9H7BrN2O
Molecular Weight (g/mol) 239.072
InChI Key AVXXUFMZRHQRTK-UHFFFAOYSA-N
PubChem CID 1214889
IUPAC Name 5-(4-bromophenyl)-3-methyl-1,2,4-oxadiazole
SMILES CC1=NOC(=N1)C2=CC=C(C=C2)Br

Specifications

Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 250 mg
Synonym 5-4-bromophenyl-3-methyl-1,2,4-oxadiazole, 1,2,4-oxadiazole,5-4-bromophenyl-3-methyl, 1,2,4-oxadiazole, 5-4-bromophenyl-3-methyl, 3-methyl-5-4-bromophenyl-1,2,4-oxadiazole, 5-4-bromo-phenyl-3-methyl-1,2,4 oxadiazole
Formula Weight 239.07
Percent Purity 97%
Chemical Name or Material 5-(4-Bromophenyl)-3-methyl-1, 2, 4-oxadiazole
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.