accessibility menu, dialog, popup

UserName

CAS RN 72-14-0

IUPAC Name: 4-amino-N-(1,3-thiazol-2-yl)benzene-1-sulfonamide
Synonyms: thiazamide planomide sulzol solfatiazolo sulphathiazole coco-thiazole poliseptil streptosilthiazole 2-sulfonamidothiazole sulfathiazole preparation
Molecular Weight (g/mol): 255.31
Molecular Formula: C9H9N3O2S2
InChi Key: JNMRHUJNCSQMMB-UHFFFAOYSA-N
SMILES: NC1=CC=C(C=C1)S(=O)(=O)NC1=NC=CS1
Molecular Weight (g/mol) 
  • (2)
  • (7)
Percent Purity 
  • (3)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
Certifications/Compliance 
  • (3)
  • (1)
Concentration 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (1)
  • (2)
  • (3)
  • (2)
  • (7)
  • (3)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (3)
  • (1)
  • (1)
  • (1)
  • (1)