accessibility menu, dialog, popup

UserName

CAS RN 7205-91-6

IUPAC Name: [(chloromethyl)sulfanyl]benzene
Synonyms: [(chloromethyl)sulfanyl]benzene
Molecular Weight (g/mol): 158.64
Molecular Formula: C7H7ClS
InChi Key: LLSMWLJPWFSMCP-UHFFFAOYSA-N
SMILES: ClCSC1=CC=CC=C1
Boiling Point 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results