missing translation for 'onlineSavingsMsg'
Learn More

Chloromethyl phenyl sulfide, 97%

Product Code. 10129499 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
25 g
Packaging:
Glass bottle
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10129499 25 g Glass bottle 25g
10017949 5 g Glass bottle 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10129499 Supplier Thermo Scientific Acros Supplier No. 202360250
 Restricted Item
Request Bulk or Custom Format
Currently Unavailable: We have no information when this product will be available.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 7205-91-6 | C7H7ClS | 158.64 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7205-91-6
Molecular Formula C7H7ClS
Molecular Weight (g/mol) 158.64
MDL Number MFCD00000921
InChI Key LLSMWLJPWFSMCP-UHFFFAOYSA-N
Synonym chloromethyl phenyl sulfide, chloromethylthiobenzene, alpha-chlorothioanisole, phenyl chloromethyl sulfide, chloromethyl phenyl sulphide, phenylthio methyl chloride, chloromethyl thio benzene, sulfide, chloromethyl phenyl, chloromethyl phenyl sulfane, benzene, chloromethyl thio
PubChem CID 81623
IUPAC Name chloromethylsulfanylbenzene
SMILES ClCSC1=CC=CC=C1

Specifications

Color Colorless to Yellow
Density 1.2300g/mL
Boiling Point 66°C (0.3 hPa)
Flash Point 101°C
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Packaging Glass bottle
Linear Formula C6H5SCH2Cl
Refractive Index 1.595
Quantity 25 g
Beilstein 06,III,1002
Fieser 09,107; 06,571; 07,357; 08,472; 09,451; 10,384; 11,503; 12,468; 13,34; 14,177; 16,98; 17,327
Specific Gravity 1.23
Formula Weight 158.65
Percent Purity 97%
Physical Form Liquid
Chemical Name or Material Chloromethyl phenyl sulfide
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.