accessibility menu, dialog, popup

UserName

CH3OOOCH3O

CAS RN 72207-94-4

IUPAC Name: 2-ethoxy-4-formylphenyl acetate
Synonyms: 2-ethoxy-4-formylphenyl acetate
Molecular Weight (g/mol): 208.21
Molecular Formula: C11H12O4
InChi Key: XRZFVPCFHPIMAD-UHFFFAOYSA-N
SMILES: CCOC1=CC(C=O)=CC=C1OC(C)=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results