accessibility menu, dialog, popup

UserName

H3NFF

CAS RN 72235-53-1

IUPAC Name: (3,4-difluorophenyl)methanaminium
Synonyms: (3,4-difluorophenyl)methanaminium
Molecular Weight (g/mol): 144.14
Molecular Formula: C7H8F2N
InChi Key: PHLZUDXEBCQHKM-UHFFFAOYSA-O
SMILES: [NH3+]CC1=CC=C(F)C(F)=C1
Molecular Weight (g/mol) 
  • (2)
  • (2)
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results