Product Code. 10013700
Shop All Thermo Scientific Chemicals Products
Click to view available options
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code.
Quantity
Packaging
Unit Size
10013700
1 g
Glass bottle
1g
10416812
5 g
Glass bottle
5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code.
10013700
Supplier
Thermo Scientific Acros
Supplier No.
311530010
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Chemical Identifiers
CAS
72235-53-1
Molecular Formula
C7 H7 F2 N
Molecular Weight (g/mol)
143.14
MDL Number
MFCD00010145
InChI Key
PHLZUDXEBCQHKM-UHFFFAOYSA-N
PubChem CID
123572
IUPAC Name
(3,4-difluorophenyl)methanamine
SMILES
C1=CC(=C(C=C1CN)F)F
Specifications
CAS Min %
96.0
CAS Max %
100.0
Color
Colorless to Yellow
Density
1.2100g/mL
Flash Point
79°C
Infrared Spectrum
Authentic
Assay Percent Range
94% min. (GC)
Packaging
Glass bottle
Linear Formula
F2 C6 H3 CH2 NH2
Refractive Index
1.4920 to 1.4940
Quantity
1 g
Specific Gravity
1.21
Synonym
3,4-difluorobenzylamine, 3,4-difluorophenyl methanamine, 3,4-difluorobenzyl amine, benzenemethanamine, 3,4-difluoro, 1-3,4-difluorophenyl methanamine, 3,4-difluorophenyl methylamine, 3, 4-difluorobenzylamine, pubchem1918, acmc-209omb, 3,4-difluoro-benzylamine
Formula Weight
143.14
Percent Purity
95%
Physical Form
Liquid
Chemical Name or Material
3, 4-Difluorobenzylamine, 97%
Show More
Show Less
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our
Privacy Policy .
Spot an opportunity for improvement?