accessibility menu, dialog, popup

UserName

CH3HO

CAS RN 7228-47-9

IUPAC Name: 1-(naphthalen-2-yl)ethan-1-ol
Synonyms: 1-(naphthalen-2-yl)ethanol
Molecular Weight (g/mol): 172.23
Molecular Formula: C12H12O
InChi Key: AXRKCRWZRKETCK-UHFFFAOYNA-N
SMILES: CC(O)C1=CC=C2C=CC=CC2=C1
Boiling Point 
  • (1)
  • (2)
Molecular Weight (g/mol) 
  • (2)
  • (1)
Percent Purity 
  • (2)
  • (1)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (2)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)