missing translation for 'onlineSavingsMsg'
Learn More

1-(2-Naphthyl)ethanol, 97%

Product Code. 15458829
Change view
Click to view available options
Quantity:
1 g
10 g
Unit Size:
10g
1g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15458829 1 g 1g
11486720 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15458829 Supplier Thermo Scientific Alfa Aesar Supplier No. L05108.03
 Restricted Item
Estimated Shipment: 04-09-2026
Add to basket
This item is not returnable. View return policy

CAS: 7228-47-9 | C12H12O | 172.227 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7228-47-9
Molecular Formula C12H12O
Molecular Weight (g/mol) 172.227
MDL Number MFCD00004111
InChI Key AXRKCRWZRKETCK-UHFFFAOYSA-N
Synonym 1-2-naphthyl ethanol, 1-naphthalen-2-yl ethanol, +/--1-2-naphthyl ethanol, 1-naphthalen-2-yl ethan-1-ol, alpha-methylnaphthalene-2-methanol, .alpha.-methyl-2-naphthalenemethanol, pubchem9427, acmc-209gxz, acmc-1azul, methyl 2-naphtylcarbinol
PubChem CID 98243
IUPAC Name 1-naphthalen-2-ylethanol
SMILES CC(C1=CC2=CC=CC=C2C=C1)O

Specifications

Melting Point 72°C to 75°C
Boiling Point 170°C (13 mmHg)
Quantity 1 g
Beilstein 1907449
Formula Weight 172.23
Percent Purity 97%
Chemical Name or Material 1-(2-Naphthyl)ethanol

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.