accessibility menu, dialog, popup

UserName

POHBrPhPhPh

CAS RN 7237-34-5

IUPAC Name: 4-methylidene-5-[2-(2-methylpropyl)-3,4-dihydro-2H-pyran-4-yl]-3-(4-nitrophenyl)-1,3-oxazolidin-2-one
Synonyms: 4-methylidene-5-[2-(2-methylpropyl)-3,4-dihydro-2H-pyran-4-yl]-3-(4-nitrophenyl)-1,3-oxazolidin-2-one
Molecular Weight (g/mol): 358.39
Molecular Formula: C19H22N2O5
InChi Key: DFQGLHYKNRBJCI-UHFFFAOYNA-N
SMILES: CC(C)CC1CC(C=CO1)C1OC(=O)N(C1=C)C1=CC=C(C=C1)[N+]([O-])=O
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results