accessibility menu, dialog, popup

UserName

OOSSOO

CAS RN 7244-02-2

IUPAC Name: 2-({2-[(carboxylatomethyl)sulfanyl]ethyl}sulfanyl)acetate
Synonyms: 2-({2-[(carboxylatomethyl)sulfanyl]ethyl}sulfanyl)acetate
Molecular Weight (g/mol): 208.25
Molecular Formula: C6H8O4S2
InChi Key: CCQMORVULNZXIA-UHFFFAOYSA-L
SMILES: [O-]C(=O)CSCCSCC([O-])=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results