missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 1,2-Bis(carboxymethylthio)ethane, 97%

Product Code. 11434527
Change view
Click to view available options
Quantity:
100 g
Unit Size:
100g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11434527 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11434527 Supplier Alfa Aesar™ Supplier No. B21074.22
Request Bulk or Custom Format

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Chemical Identifiers

CAS 7244-02-2

Specifications

Melting Point 110°C
Molecular Formula C31H24BrP
MDL Number MFCD00031650
Quantity 100 g
Synonym 9-fluorenyl triphenylphosphonium bromide, 9h-fluoren-9-yltriphenylphosphanium bromide, 9-fluorenyltriphenylphosphium bromide, 9h-fluoren-9-yl triphenylphosphonium bromide
InChI Key BHNPKGAMAZKCCJ-UHFFFAOYSA-M
SMILES [Br-].C1=CC=C(C=C1)[P+](C1C2=CC=CC=C2C2=CC=CC=C12)(C1=CC=CC=C1)C1=CC=CC=C1
IUPAC Name 9H-fluoren-9-yl(triphenyl)phosphanium;bromide
Molecular Weight (g/mol) 507.41
PubChem CID 18702719
Formula Weight 210.26
Percent Purity 97%
Chemical Name or Material 1,2-Bis(carboxymethylthio)ethane
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.