accessibility menu, dialog, popup

UserName

CAS RN 72599-27-0

H3CN(S)OH(R)OH(R)HO(R)OH

CAS RN 72599-27-0

IUPAC Name: (2R,3R,4R,5S)-1-butyl-2-(hydroxymethyl)piperidine-3,4,5-triol
Synonyms: miglustat butyldeoxynojirimycin n-butyl dnj zavesca BuDNJ N-butyl-1-deoxynojirimycin miglustatum N-butylmoranoline NB-dnj n-butyldeoxynojirimycin
Molecular Weight (g/mol): 219.28
Molecular Formula: C10H21NO4
InChi Key: UQRORFVVSGFNRO-UTINFBMNSA-N
SMILES: CCCCN1C[C@H](O)[C@@H](O)[C@H](O)[C@H]1CO

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results