accessibility menu, dialog, popup

UserName

CAS RN 728-80-3

FFFHO

CAS RN 728-80-3

IUPAC Name: phenyl[3-(trifluoromethyl)phenyl]methanol
Synonyms: phenyl[3-(trifluoromethyl)phenyl]methanol
Molecular Weight (g/mol): 252.24
Molecular Formula: C14H11F3O
InChi Key: KRMIEGLFYDWJOT-UHFFFAOYNA-N
SMILES: OC(C1=CC=CC=C1)C1=CC(=CC=C1)C(F)(F)F
Boiling Point 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results