CAS RN 73200-73-4
CAS RN 73200-73-4
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
2
of
2
results
1
3-Phenylpyrazin-2-ol, Mikromol™
Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.
2
3-Phenylpyrazin-2-ol, TRC
CAS: 73200-73-4 Molecular Formula: C10 H8 N2 O Molecular Weight (g/mol): 172.18 Synonym: 3-Phenylpyrazin-2-ol,Cefaclor Imp. F (EP),Ampicillin Sodium Imp. H (EP) IUPAC Name: 3-phenylpyrazin-2-ol SMILES: Oc1nccnc1c2ccccc2