accessibility menu, dialog, popup

UserName

OHNH2

CAS RN 73604-31-6

IUPAC Name: 3-(aminomethyl)phenol
Synonyms: 3-(aminomethyl)phenol
Molecular Weight (g/mol): 123.16
Molecular Formula: C7H9NO
InChi Key: JNZYADHPGVZMQK-UHFFFAOYSA-N
SMILES: NCC1=CC(O)=CC=C1
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results