missing translation for 'onlineSavingsMsg'
Learn More

3-Hydroxybenzylamine, 96%, Thermo Scientific Chemicals

Product Code. 15467557
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
Unit Size:
1g
250mg
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15467557 1 g 1g
15487557 250 mg 250mg
15477557 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 15467557 Supplier Thermo Scientific Alfa Aesar Supplier No. H35708.03
 Restricted Item
Estimated Shipment: 30-09-2026
Add to basket
This item is not returnable. View return policy

3-Hydroxybenzylamine is used as pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
3-Hydroxybenzylamine is used as pharmaceutical intermediate.

Solubility
Soluble in water and ethanol.

Notes
Air sensitive. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 73604-31-6
Molecular Formula C7H9NO
Molecular Weight (g/mol) 123.16
MDL Number MFCD00798977
InChI Key JNZYADHPGVZMQK-UHFFFAOYSA-N
Synonym 3-aminomethyl phenol, 3-hydroxybenzylamine, phenol, 3-aminomethyl, 3-hydroxybenzylamine, 3-hydroxyphenyl methylamine, 3-aminomethyl-phenol, 3-hydroxy-benzylamine, 3-hydroxyphenyl methylamine
PubChem CID 735894
IUPAC Name 3-(aminomethyl)phenol
SMILES NCC1=CC(O)=CC=C1

Specifications

Melting Point 170°C to 173°C
Quantity 1 g
UN Number UN3259
Sensitivity Air sensitive
Solubility Information Soluble in water and ethanol.
Formula Weight 123.15
Percent Purity 97%
Chemical Name or Material 3-Hydroxybenzylamine

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.