accessibility menu, dialog, popup

UserName

CAS RN 7365-44-8

IUPAC Name: 2-{[1,3-dihydroxy-2-(hydroxymethyl)propan-2-yl]amino}ethane-1-sulfonic acid
Synonyms: tes (buffer)
Molecular Weight (g/mol): 229.25
Molecular Formula: C6H15NO6S
InChi Key: JOCBASBOOFNAJA-UHFFFAOYSA-N
SMILES: OCC(CO)(CO)NCCS(O)(=O)=O
Quantity 
  • (1)
  • (4)
  • (1)
  • (1)
  • (1)
  • (1)
Percent Purity 
  • (1)
  • (5)
Physical Form 
  • (2)
  • (1)
  • (1)
pH 
  • (2)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (2)
  • (2)
  • (2)
  • (1)
  • (4)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (5)
  • (2)
  • (1)
  • (1)
  • (2)
  • (1)
  • (2)