missing translation for 'onlineSavingsMsg'
Learn More

TES (White Crystals), Fisher BioReagents

Product Code. 10387023
Change view
Click to view available options
Quantity:
100 g
Unit Size:
100g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
10387023 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10387023 Supplier Fisher BioReagents Supplier No. BP309100
Request Bulk or Custom Format

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy
This biological buffer has a usable pH range of 6.8 to 8.2.
TRUSTED_SUSTAINABILITY

Specifications

CAS 7365-44-8
Melting Point 223°C
Color White
Decomposition Information >220°C
pH 4.0 to 4.8
Physical Form Powder or Solid
Quantity 100 g
Assay Percent Range ≥98 %
Identification Pass Test
Molecular Formula C6H15NO6S
Packaging Glass Bottle
Merck Index 15, 9319
Vapor Pressure Negligible
Synonym N-Tris(hydroxymethyl)methyl-2-aminoethanesulfonic Acid
InChI Key JOCBASBOOFNAJA-UHFFFAOYSA-N
SMILES C(CS(=O)(=O)O)NC(CO)(CO)CO
IUPAC Name 2-[[1,3-dihydroxy-2-(hydroxymethyl)propan-2-yl]amino]ethanesulfonic acid
Molecular Weight (g/mol) 229.247
PubChem CID 81831
ChEBI CHEBI:44356
Formula Weight 229.25
Percent Purity ≥98%
Chemical Name or Material TES
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.