accessibility menu, dialog, popup

UserName

H2NPH2NOO

CAS RN 7450-69-3

IUPAC Name: [(diaminophosphoryl)oxy]benzene
Synonyms: [(diaminophosphoryl)oxy]benzene
Molecular Weight (g/mol): 172.12
Molecular Formula: C6H9N2O2P
InChi Key: AYRRNFHDJUXLEQ-UHFFFAOYSA-N
SMILES: NP(N)(=O)OC1=CC=CC=C1
Molecular Weight (g/mol) 
  • (1)
  • (2)
Percent Purity 
  • (2)
  • (1)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (1)
  • (2)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)