missing translation for 'onlineSavingsMsg'
Learn More

Phenyl phosphorodiamidate, 97%, Thermo Scientific™

Product Code. 10763521 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
5g; Glass bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code.
10763521 5g; Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10763521 Supplier Thermo Scientific Acros Supplier No. 300440050

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Specifications

CAS 7450-69-3
Infrared Spectrum Authentic
Assay Percent Range 97%
Packaging Glass bottle
Molecular Formula C6H9N2O2P
MDL Number MFCD00014767
Quantity 5g
Beilstein 06, IV, 751
Synonym phenyl phosphorodiamidate, phenylphosphorodiamidate, phenyl diamidophosphate, phosphorodiamidic acid, phenyl ester, diaminophosphoryl oxy benzene, phenylphosphorodiamidat, phosphoric phenyl ester diamide, phenyl phosphordiamidate, phenyl diamidophosphate #, diaminophenoxyphosphino-1-one
Solubility Information Solubility in water: soluble. Other solubilities: soluble in methanol,ethanol,chcl3,ccl4
InChI Key AYRRNFHDJUXLEQ-UHFFFAOYSA-N
SMILES C1=CC=C(C=C1)OP(=O)(N)N
IUPAC Name diaminophosphoryloxybenzene
Molecular Weight (g/mol) 172.12
PubChem CID 81954
Formula Weight 172.12
Percent Purity 97%
Chemical Name or Material Phenyl phosphorodiamidate, 97%
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.