accessibility menu, dialog, popup

UserName

CH3SiCH3CH3H3C

CAS RN 75-76-3

IUPAC Name: tetramethylsilane
Synonyms: tetramethylsilane
Molecular Weight (g/mol): 88.23
Molecular Formula: C4H12Si
InChi Key: CZDYPVPMEAXLPK-UHFFFAOYSA-N
SMILES: C[Si](C)(C)C
Quantity 
  • (3)
  • (2)
  • (3)
  • (1)
Boiling Point 
  • (3)
  • (4)
Percent Purity 
  • (3)
  • (2)
  • (2)
Physical Form 
  • (5)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (2)
  • (3)
  • (1)
  • (3)
  • (4)
  • (3)
  • (2)
  • (2)
  • (5)
  • (2)
  • (2)