missing translation for 'onlineSavingsMsg'
Learn More

Tetramethylsilane, 99.9+%, NMR grade

Product Code. 10615192 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
25 g
100 g
Packaging:
Glass bottle
Unit Size:
100g
25g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10615192 25 g Glass bottle 25g
10553301 100 g Glass bottle 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10615192 Supplier Thermo Scientific Acros Supplier No. 138470250
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 75-76-3 | C4H12Si | 88.23 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

For NMR
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 75-76-3
Molecular Formula C4H12Si
Molecular Weight (g/mol) 88.23
MDL Number MFCD00008274
InChI Key CZDYPVPMEAXLPK-UHFFFAOYSA-N
Synonym silane, tetramethyl, tetramethylsilicane, tetramethyl silane, silicon, tetramethyl, tetramethyl-silane, unii-41y0rbg14q, me4si, ch3 4si, si ch3 4, chembl68073
PubChem CID 6396
ChEBI CHEBI:85361
IUPAC Name tetramethylsilane
SMILES C[Si](C)(C)C

Specifications

Melting Point -99.0°C
Color Colorless
Density 0.6400g/mL
Boiling Point 26.0°C to 28.0°C
Flash Point −27°C
Assay Percent Range 99.9% min. (GC)
Packaging Glass bottle
Linear Formula Si(CH3)4
Refractive Index 1.3585
Quantity 25 g
Beilstein 04, 625
Merck Index 15, 9377
Specific Gravity 0.64
Solubility Information Solubility in water: 0.02g/L (25°C). Other solubilities: soluble in most common organic solvents
Formula Weight 88.22
Percent Purity 99.9+%
Physical Form Volatile Liquid
Chemical Name or Material Tetramethylsilane, 99.9+%
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.