accessibility menu, dialog, popup

UserName

CAS RN 7524-52-9

NH(S)ONH2OCH3HCl

CAS RN 7524-52-9

IUPAC Name: (2S)-3-(1H-indol-3-yl)-1-methoxy-1-oxopropan-2-aminium
Synonyms: (2S)-3-(1H-indol-3-yl)-1-methoxy-1-oxopropan-2-aminium
Molecular Weight (g/mol): 219.26
Molecular Formula: C12H15N2O2
InChi Key: KCUNTYMNJVXYKZ-JTQLQIEISA-O
SMILES: COC(=O)[C@@H]([NH3+])CC1=CNC2=CC=CC=C12

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results