accessibility menu, dialog, popup

UserName

CAS RN 75708-92-8

ONHOOHOHONHNNONHNH2NH2OH2O

CAS RN 75708-92-8

IUPAC Name: 2-[(4-{[(2-amino-4-oxo-1,4-dihydropteridin-6-yl)methyl]amino}phenyl)formamido]pentanedioic acid dihydrate
Synonyms: 2-[(4-{[(2-amino-4-oxo-1H-pteridin-6-yl)methyl]amino}phenyl)formamido]pentanedioic acid dihydrate
Molecular Weight (g/mol): 477.43
Molecular Formula: C19H23N7O8
InChi Key: ODYNNYOEHBJUQP-UHFFFAOYNA-N
SMILES: O.O.NC1=NC(=O)C2=NC(CNC3=CC=C(C=C3)C(=O)NC(CCC(O)=O)C(O)=O)=CN=C2N1
Molecular Weight (g/mol) 
  • (1)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results