missing translation for 'onlineSavingsMsg'
Learn More

Folic acid dihydrate, 97%

Product Code. 11492477
Change view
Click to view available options
Quantity:
25 g
100 g
Unit Size:
100g
25g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11492477 100 g 100g
11482477 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11492477 Supplier Thermo Scientific Alfa Aesar Supplier No. A14300.22
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

CAS: 75708-92-8 | C19H23N7O8 | 477.434 g/mol

Vitamin B9. A B-complex vitamin necessary for the body to produce red blood cellsFolic acid dehydrate is used to form non-covalent complexes with polyethylenimine. It is used for the delivery of genes, which finds application in gene therapy. It is a B-complex vitamin, which is necessary for the body to produce red blood cells.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Vitamin B9. A B-complex vitamin necessary for the body to produce red blood cellsFolic acid dehydrate is used to form non-covalent complexes with polyethylenimine. It is used for the delivery of genes, which finds application in gene therapy. It is a B-complex vitamin, which is necessary for the body to produce red blood cells.

Solubility
Miscible with water.

Notes
Light sensitive. Incompatible with strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 75708-92-8
Molecular Formula C19H23N7O8
Molecular Weight (g/mol) 477.434
MDL Number MFCD00079305
InChI Key ODYNNYOEHBJUQP-LTCKWSDVSA-N
Synonym folic acid dihydrate, unii-0s1t5c0jzl, 0s1t5c0jzl, pubchem18447, l-glutamic acid, n-4-2-amino-1,4-dihydro-4-oxo-6-pteridinyl methyl amino benzoyl-, dihydrate, l-glutamic acid, n-4-2-amino-3,4-dihydro-4-oxo-6-pteridinyl methyl amino benzoyl-, hydrate 1:2, folicacid,vitaminmorvitaminbc,vitaminm,pteglu, 2s-2-4-2-amino-4-hydroxypteridin-6-yl methyl amino phenyl formamido pentanedioic acid dihydrate, s-2-4-2-amino-4-oxo-3,4-dihydropteridin-6-yl methyl amino benzamido pentanedioic acid dihydrate
PubChem CID 16211651
IUPAC Name (2S)-2-[[4-[(2-amino-4-oxo-1H-pteridin-6-yl)methylamino]benzoyl]amino]pentanedioic acid;dihydrate
SMILES C1=CC(=CC=C1C(=O)NC(CCC(=O)O)C(=O)O)NCC2=CN=C3C(=N2)C(=O)N=C(N3)N.O.O

Specifications

Melting Point ∼250°C (decomposition)
Quantity 100 g
Beilstein 100781
Merck Index 14,4221
Solubility Information Miscible with water.
Optical Rotation +18° (c=0.5 in 0.1N NaOH)
Formula Weight 477.44 (441.40 Anhydrous)
Percent Purity 97%
Chemical Name or Material Folic acid dihydrate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.