accessibility menu, dialog, popup

UserName

CAS RN 76666-32-5

H3COOOHO

CAS RN 76666-32-5

IUPAC Name: 3-hydroxy-2-(3-methoxyphenyl)-4H-chromen-4-one
Synonyms: 3-hydroxy-2-(3-methoxyphenyl)chromen-4-one
Molecular Weight (g/mol): 268.27
Molecular Formula: C16H12O4
InChi Key: GYLGASXCHFNKHD-UHFFFAOYSA-N
SMILES: COC1=CC=CC(=C1)C1=C(O)C(=O)C2=CC=CC=C2O1
Molecular Weight (g/mol) 
  • (1)
  • (3)
Percent Purity 
  • (3)
  • (1)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
Grade 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results