accessibility menu, dialog, popup

UserName

CAS RN 76721-89-6

HOONHOSH

CAS RN 76721-89-6

IUPAC Name: 2-(2-benzyl-3-sulfanylpropanamido)acetic acid
Synonyms: thiorphan
Molecular Weight (g/mol): 253.32
Molecular Formula: C12H15NO3S
InChi Key: LJJKNPQAGWVLDQ-UHFFFAOYNA-N
SMILES: OC(=O)CNC(=O)C(CS)CC1=CC=CC=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)