accessibility menu, dialog, popup

UserName

H3CH3CCH3CH3OHHOOHHO

CAS RN 77-08-7

IUPAC Name: 3,3,3',3'-tetramethyl-2,2',3,3'-tetrahydro-1,1'-spirobi[indene]-5,5',6,6'-tetrol
Synonyms: 3,3,3',3'-tetramethyl-2H,2'H-1,1'-spirobi[indene]-5,5',6,6'-tetrol
Molecular Weight (g/mol): 340.42
Molecular Formula: C21H24O4
InChi Key: POFMQEVZKZVAPQ-UHFFFAOYSA-N
SMILES: CC1(C)CC2(CC(C)(C)C3=CC(O)=C(O)C=C23)C2=CC(O)=C(O)C=C12

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results