missing translation for 'onlineSavingsMsg'
Learn More

5,5',6,6'-Tetrahydroxy-3,3,3',3'-tetramethyl-1,1'-spirobisindane, 97%

Product Code. 11479697
Change view
Click to view available options
Quantity:
50 g
250 g
Unit Size:
250g
50g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11479697 50 g 50g
11489697 250 g 250g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11479697 Supplier Thermo Scientific Alfa Aesar Supplier No. B22170.18
 Restricted Item
Estimated Shipment: 09-11-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 77-08-7 | C21H24O4 | 340.419 g/mol

5,5',6,6'-Tetrahydroxy-3,3,3',3'- tetramethyl-1,1'-spirobisindane is used in the preparation of 4,7,4',7'-tetrabromo-3,3,3',3'-tetramethyl-2,3,2',3'-tetrahydro-[1,1']spirobiindene-5,6,5',6'-tetraol by bromination using bromine.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
5,5′,6,6′-Tetrahydroxy-3,3,3′,3′- tetramethyl-1,1′-spirobisindane is used in the preparation of 4,7,4′,7′-tetrabromo-3,3,3′,3′-tetramethyl-2,3,2′,3′-tetrahydro-[1,1′]spirobiindene-5,6,5′,6′-tetraol by bromination using bromine.

Solubility
Slightly soluble in methanol.

Notes
Incompatible with strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 77-08-7
Molecular Formula C21H24O4
Molecular Weight (g/mol) 340.419
MDL Number MFCD00021235
InChI Key POFMQEVZKZVAPQ-UHFFFAOYSA-N
Synonym 5,5',6,6'-tetrahydroxy-3,3,3',3'-tetramethyl-1,1'-spirobisindane, tts 5, 5,5',6,6'-tetrahydroxy-3,3,3',3'-tetramethyl-1,1'-spirobiindan, 1,1'-spirobi 1h-indene-5,5',6,6'-tetrol, 2,2',3,3'-tetrahydro-3,3,3',3'-tetramethyl, 1,1'-spirobi indane-5,6-diol , 3,3,3',3'-tetramethyl, 1,1,1',1'-tetramethyl-3,3'-spirobi 2h-indene-5,5',6,6'-tetrol, 3,3,3',3'-tetramethyl-2h,2'h-1,1'-spirobi indene-5,5',6,6'-tetrol, 3,3,3',3'-tetramethyl-2,2',3,3'-tetrahydro-1,1'-spirobi indene-5,5',6,6'-tetrol, 1,6-diol , 3,3,3',3'-tetramethyl
PubChem CID 66162
IUPAC Name 1,1,1',1'-tetramethyl-3,3'-spirobi[2H-indene]-5,5',6,6'-tetrol
SMILES CC1(CC2(CC(C3=CC(=C(C=C32)O)O)(C)C)C4=CC(=C(C=C41)O)O)C

Specifications

Melting Point >300°C
Quantity 50 g
Beilstein 2226231
Solubility Information Slightly soluble in methanol.
Formula Weight 340.42
Percent Purity 97%
Chemical Name or Material 5,5',6,6'-Tetrahydroxy-3,3,3',3'-tetramethyl-1,1'-spirobisindane
compliance-icons
Product Identifier
  • 5,5',6,6'-Tetrahydroxy-3,3,3',3'-tetramethyl-1,1'-spirobisindane
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
  • P332+P313-If skin irritation occurs: Get medical advice/attention.
  • P337+P313-If eye irritation persists: Get medical advice/attention.
Supplemental information
  • MIXTURE LIST-Contain: 2,2',3,3'-tetrahydro-3,3,3',3'-tetramethyl-1,1'-Spirobi[1H-indene]-5,5',6,6'-tetrol

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.