accessibility menu, dialog, popup

UserName

CAS RN 77-77-0

CH2SOOH2C

CAS RN 77-77-0

IUPAC Name: (ethenesulfonyl)ethene
Synonyms: vinyl sulfone
Molecular Weight (g/mol): 118.15
Molecular Formula: C4H6O2S
InChi Key: AFOSIXZFDONLBT-UHFFFAOYSA-N
SMILES: C=CS(=O)(=O)C=C
Molecular Weight (g/mol) 
  • (2)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
Boiling Point 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)