missing translation for 'onlineSavingsMsg'
Learn More

Divinyl sulfone, 97%, stab. with 0.05% hydroquinone

Product Code. 11383386
Change view
Click to view available options
Quantity:
10 g
50 g
Unit Size:
10g
50g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11383386 10 g 10g
11393386 50 g 50g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11383386 Supplier Thermo Scientific Alfa Aesar Supplier No. L12827.09
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

CAS: 77-77-0 | C4H6O2S | 118.15 g/mol

Divinyl sulfone acts as a cross-linking reagent for agarose gels. It is a monomer, which is used in the production of polymers with diols, urea and malonic esters. Further, it serves as a shrinkage control agent in textiles. In addition to this, it is used in the preparation of large class of fiber-reactive dyestuffs.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Divinyl sulfone acts as a cross-linking reagent for agarose gels. It is a monomer, which is used in the production of polymers with diols, urea and malonic esters. Further, it serves as a shrinkage control agent in textiles. In addition to this, it is used in the preparation of large class of fiber-reactive dyestuffs.

Solubility
Miscible with water.

Notes
Incompatible with strong oxidizing agents, strong acids, reducing agents, alkali metals and strong bases.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 77-77-0
Molecular Formula C4H6O2S
Molecular Weight (g/mol) 118.15
MDL Number MFCD00008623
InChI Key AFOSIXZFDONLBT-UHFFFAOYSA-N
Synonym divinyl sulfone, vinyl sulfone, divinyl sulphone, bis ethenyl sulfone, vinyl sulphone, sulfone, divinyl, ethenesulfonyl-ethene, ethene, 1,1'-sulfonylbis, 1,1'-sulphonylbisethene, ethenesulfonyl ethene
PubChem CID 6496
ChEBI CHEBI:53729
IUPAC Name 1-ethenylsulfonylethene
SMILES C=CS(=O)(=O)C=C

Specifications

Melting Point ∼-26°C
Density 1.177
Boiling Point 233°C to 234°C
Flash Point 102°C (215°F)
Refractive Index 1.476
Quantity 10 g
UN Number UN2927
Beilstein 1071329
Solubility Information Miscible with water.
Formula Weight 118.16
Percent Purity 97%
Chemical Name or Material Divinyl sulfone, stabilized with 0.05% hydroquinone
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • Divinyl sulfone
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 1
  • Acute toxicity Category 2
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H300-Fatal if swallowed.
  • H310-Fatal in contact with skin.
  • H314-Causes severe skin burns and eye damage.
  • H318-Causes serious eye damage.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
  • P330-Rinse mouth.
Supplemental information
  • MIXTURE LIST-Contain: Divinyl sulphone

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.