accessibility menu, dialog, popup

UserName

CAS RN 771-98-2

IUPAC Name: (cyclohex-1-en-1-yl)benzene
Synonyms: 1-phenylcyclohexene
Molecular Weight (g/mol): 158.24
Molecular Formula: C12H14
InChi Key: WCMSFBRREKZZFL-UHFFFAOYSA-N
SMILES: C1CCC(=CC1)C1=CC=CC=C1
Molecular Weight (g/mol) 
  • (5)
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (2)
Boiling Point 
  • (3)
  • (2)
Percent Purity 
  • (2)
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (5)
  • (3)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (2)
  • (3)
  • (2)
  • (2)
  • (3)
  • (2)