missing translation for 'onlineSavingsMsg'
Learn More

1-Phenylcyclohexene, 96%

Product Code. 11459017
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11459017 100 g 100g
11439017 5 g 5g
11449017 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11459017 Supplier Thermo Scientific Alfa Aesar Supplier No. B20602.22
 Restricted Item
Estimated Shipment: 04-09-2026
Add to basket
This item is not returnable. View return policy

CAS: 771-98-2 | C12H14 | 158.244 g/mol

It is used as a pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is used as a pharmaceutical intermediate.

Solubility
Insoluble in water.

Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 771-98-2
Molecular Formula C12H14
Molecular Weight (g/mol) 158.244
MDL Number MFCD00001542
InChI Key WCMSFBRREKZZFL-UHFFFAOYSA-N
Synonym 1-phenyl-1-cyclohexene, 1-phenylcyclohexene, 1-phenylcyclohex-1-ene, benzene, 1-cyclohexen-1-yl, phenylcyclohexene, cyclohexenylbenzene, benzene, cyclohexenyl, 2,3,4,5-tetrahydro-1,1'-biphenyl, cyclohexene, 1-phenyl, trans-1-phenylcyclohexene
PubChem CID 13043
IUPAC Name cyclohexen-1-ylbenzene
SMILES C1CCC(=CC1)C2=CC=CC=C2

Specifications

Melting Point -11°C
Density 0.992
Boiling Point 250°C to 252°C
Flash Point 103°C (217°F)
Refractive Index 1.57
Quantity 100 g
Beilstein 1905772
Solubility Information Insoluble in water.
Formula Weight 158.24
Percent Purity 97%
Chemical Name or Material 1-Phenylcyclohexene
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.