missing translation for 'onlineSavingsMsg'
Learn More

1-Phenyl-1-cyclohexene, 95%, Thermo Scientific Chemicals

Product Code. 10356630 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
25 g
Packaging:
Glass bottle
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10356630 25 g Glass bottle 25g
10255130 5 g Glass bottle 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10356630 Supplier Thermo Scientific Acros Supplier No. 130450250
Request Bulk or Custom Format

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 771-98-2
Molecular Formula C12H14
Molecular Weight (g/mol) 158.24
MDL Number MFCD00001542
InChI Key WCMSFBRREKZZFL-UHFFFAOYSA-N
Synonym 1-phenyl-1-cyclohexene, 1-phenylcyclohexene, 1-phenylcyclohex-1-ene, benzene, 1-cyclohexen-1-yl, phenylcyclohexene, cyclohexenylbenzene, benzene, cyclohexenyl, 2,3,4,5-tetrahydro-1,1'-biphenyl, cyclohexene, 1-phenyl, trans-1-phenylcyclohexene
PubChem CID 13043
IUPAC Name cyclohexen-1-ylbenzene
SMILES C1CCC(=CC1)C2=CC=CC=C2

Specifications

CAS Min % 94.0
CAS Max % 100.0
Melting Point -11°C
Color Colorless to Yellow
Density 0.9900g/mL
Boiling Point 251°C to 253°C
Flash Point 103°C
Infrared Spectrum Authentic
Assay Percent Range 94% min. (GC)
Packaging Glass bottle
Linear Formula C6H5C6H9
Refractive Index 1.5680 to 1.57
Quantity 25 g
Beilstein 05, II, 419
Specific Gravity 0.99
Formula Weight 158.24
Percent Purity 95%
Physical Form Liquid
Chemical Name or Material 1-Phenyl-1-cyclohexene, 95%
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.