CAS RN 7719-09-7
CAS RN 7719-09-7
IUPAC Name:
sulfurooyl dichloride
Synonyms:
thionyl chloride socl2 sulfinyl chloride thionyl dichloride sulfurous oxychloride sulfur oxychloride sulfur chloride oxide sulfinyl dichloride sulfur dichloride oxide
Molecular Weight (g/mol):
118.96
Molecular Formula:
Cl2OS
InChi Key:
FYSNRJHAOHDILO-UHFFFAOYSA-N
SMILES:
ClS(Cl)=O
1
–
3
of
3
results
1
Promotions Available
Thionyl chloride, 99.7%, Thermo Scientific Chemicals
Thionyl chloride, 99.7%, Thermo Scientific Chemicals
CAS: 7719-09-7 Molecular Formula: Cl2OS Molecular Weight (g/mol): 118.96 InChI Key: FYSNRJHAOHDILO-UHFFFAOYSA-N IUPAC Name: sulfurooyl dichloride SMILES: ClS(Cl)=O
2
Thionyl chloride, 1M soln. in dichloromethane, Thermo Scientific Chemicals
CAS: 7719-09-7 MDL Number: MFCD00011449
3
Thionyl chloride, 99.5+%, Thermo Scientific Chemicals
CAS: 7719-09-7 MDL Number: MFCD00011449 PubChem CID: 24386 ChEBI: CHEBI:29290