missing translation for 'onlineSavingsMsg'
Learn More

Thionyl chloride, 1M soln. in dichloromethane, Thermo Scientific Chemicals

Product Code. 15494507
Change view
Click to view available options
Quantity:
100 mL
500 mL
Unit Size:
100mL
500mL
Product Code. Quantity unitSize
15494507 100 mL 100mL
15404517 500 mL 500mL
2 options
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

Product Code. 15494507

Brand: Thermo Scientific Alfa Aesar H31857.AE

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

Sulfuryl chloride and thionyl chloride have been found to be new chlorinating agents for alkyl sulfides. Preparation of aromatic polyesters by direct polycondensation with thionyl chloride in pyridine. Conversion of aromatic and. alpha.,. beta.-unsaturated aldehydes to dichlorides by thionyl chloride and dimethylformamide.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7719-09-7
MDL Number MFCD00011449

Specifications

Molecular Formula Cl3P
Linear Formula SOCl2
UN Number UN2922
Concentration 1M soln. in dichloromethane
Density 1.35
Synonym phosphorus trichloride, phosphorous trichloride, trichlorophosphine, phosphorus chloride, phosphorous chloride, phosphorus iii chloride, pcl3, phosphorus chloride pcl3, phosphine, trichloro, fosfortrichloride
Solubility Information Reacts with water.
InChI Key FAIAAWCVCHQXDN-UHFFFAOYSA-N
SMILES P(Cl)(Cl)Cl
IUPAC Name trichlorophosphane
Molecular Weight (g/mol) 137.32
PubChem CID 24387
ChEBI CHEBI:30334
Formula Weight 118.97
Quantity 100 mL
Sensitivity Moisture sensitive; May form a precipitate
Chemical Name or Material Thionyl chloride
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.