accessibility menu, dialog, popup

UserName

CH3CH3H3CSClOO

CAS RN 773-64-8

IUPAC Name: 2,4,6-trimethylbenzene-1-sulfonyl chloride
Synonyms: 2,4,6-trimethylbenzenesulfonyl chloride
Molecular Weight (g/mol): 218.7
Molecular Formula: C9H11ClO2S
InChi Key: PVJZBZSCGJAWNG-UHFFFAOYSA-N
SMILES: CC1=CC(C)=C(C(C)=C1)S(Cl)(=O)=O
Boiling Point 
  • (3)
  • (3)
Molecular Weight (g/mol) 
  • (3)
  • (3)
Quantity 
  • (2)
  • (2)
  • (2)
Physical Form 
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (3)
  • (3)
  • (3)
  • (2)
  • (2)
  • (2)
  • (3)