missing translation for 'onlineSavingsMsg'
Learn More

Mesitylene-2-sulfonyl chloride, 99%

Product Code. 11499343
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
Unit Size:
100g
25g
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11499343 500 g 500g
11479343 25 g 25g
11489343 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11499343 Supplier Thermo Scientific Alfa Aesar Supplier No. A11775.36
 Restricted Item
Estimated Shipment: 13-11-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 773-64-8 | C9H11ClO2S | 218.695 g/mol

Mesitylene-2-sulfonyl chloride is used as reagent for intramolecular lactamizations and lactonization. It also acts as a reagent for protecting guanidino groups and tryptophan. It is used as coupling reagent in polynucleotide synthesis

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Mesitylene-2-sulfonyl chloride is used as reagent for intramolecular lactamizations and lactonization. It also acts as a reagent for protecting guanidino groups and tryptophan. It is used as coupling reagent in polynucleotide synthesis

Solubility
Soluble in toluene (almost transparency).

Notes
Moisture Sensitive. Store under inert gas. Incompatible with strong bases. Store away from moisture.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 773-64-8
Molecular Formula C9H11ClO2S
Molecular Weight (g/mol) 218.695
MDL Number MFCD00007434
InChI Key PVJZBZSCGJAWNG-UHFFFAOYSA-N
Synonym 2-mesitylenesulfonyl chloride, mesitylenesulfonyl chloride, 2,4,6-trimethylbenzene-1-sulfonyl chloride, benzenesulfonyl chloride, 2,4,6-trimethyl, mesitylsulfonyl chloride, 2-mesitylenesulfonylchloride, mesitylene-2-sulphonyl chloride, 2,4,6-trimethyl-benzenesulfonyl chloride, 2,4,6-trimethylphenylsulfonyl chloride, mesitylsulfonylchloride
PubChem CID 13046
IUPAC Name 2,4,6-trimethylbenzenesulfonyl chloride
SMILES CC1=CC(=C(C(=C1)C)S(=O)(=O)Cl)C

Specifications

Melting Point 53°C to 57°C
Boiling Point 150°C (20 mmHg)
Flash Point >110°C (230°F)
Odor Irritating
Quantity 500 g
UN Number UN3261
Beilstein 1107601
Sensitivity Moisture sensitive
Solubility Information Soluble in toluene (almost transparency).
Formula Weight 218.7
Percent Purity 99%
Chemical Name or Material Mesitylene-2-sulfonyl chloride
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.