missing translation for 'onlineSavingsMsg'
Learn More

2-Mesitylenesulfonyl chloride, 99%

Product Code. 10081270 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
Packaging:
Plastic bottle
Unit Size:
100g
25g
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10081270 25 g Plastic bottle 25g
10376810 100 g Plastic bottle 100g
10406750 500 g Plastic bottle 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10081270 Supplier Thermo Scientific Acros Supplier No. 125590250
 Restricted Item
Estimated Shipment: 11-11-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 773-64-8 | C9H11ClO2S | 218.7 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 773-64-8
Molecular Formula C9H11ClO2S
Molecular Weight (g/mol) 218.7
MDL Number MFCD00007434
InChI Key PVJZBZSCGJAWNG-UHFFFAOYSA-N
Synonym 2-mesitylenesulfonyl chloride, mesitylenesulfonyl chloride, 2,4,6-trimethylbenzene-1-sulfonyl chloride, benzenesulfonyl chloride, 2,4,6-trimethyl, mesitylsulfonyl chloride, 2-mesitylenesulfonylchloride, mesitylene-2-sulphonyl chloride, 2,4,6-trimethyl-benzenesulfonyl chloride, 2,4,6-trimethylphenylsulfonyl chloride, mesitylsulfonylchloride
PubChem CID 13046
IUPAC Name 2,4,6-trimethylbenzenesulfonyl chloride
SMILES CC1=CC(=C(C(=C1)C)S(=O)(=O)Cl)C

Specifications

CAS Min % 98.5
CAS Max % 100.0
Melting Point 53°C to 57°C
Color Gray to White
Boiling Point 150°C (20.0 mmHg)
Flash Point >110°C
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (ex Cl) (Argentometry)
Packaging Plastic bottle
Linear Formula (CH3)3C6H2SO2Cl
Quantity 25 g
Beilstein 11, 136
Fieser 01,661; 05,430; 06,625; 08,318
Solubility Information Solubility in water: reacts. Other solubilities: soluble in ether,toluene and dichloromethane
Formula Weight 218.7
Percent Purity 99%
Physical Form Crystalline Powder
Chemical Name or Material 2-Mesitylenesulfonyl chloride, 99%
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.