accessibility menu, dialog, popup

UserName

ONONFFF

CAS RN 778-94-9

IUPAC Name: 2-nitro-4-(trifluoromethyl)benzonitrile
Synonyms: 2-nitro-4-(trifluoromethyl)benzonitrile
Molecular Weight (g/mol): 216.12
Molecular Formula: C8H3F3N2O2
InChi Key: BQCWLXXZTCLGSZ-UHFFFAOYSA-N
SMILES: [O-][N+](=O)C1=CC(=CC=C1C#N)C(F)(F)F
Boiling Point 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results