missing translation for 'onlineSavingsMsg'
Learn More

2-Nitro-4-(trifluoromethyl)benzonitrile, 98%, Thermo Scientific™

Product Code. 11368505
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11368505 25 g 25g
11358505 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11368505 Supplier Thermo Scientific Alfa Aesar Supplier No. L10557.14
Request Bulk or Custom Format

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 778-94-9
Molecular Formula C8H3F3N2O2
Molecular Weight (g/mol) 216.12
MDL Number MFCD00014684
InChI Key BQCWLXXZTCLGSZ-UHFFFAOYSA-N
Synonym 2-nitro-4-trifluoromethyl benzonitrile, 4-cyano-3-nitrobenzotrifluoride, unii-e6z0mkd079, 2-nitro-4-trifluoromethyl-benzonitrile, benzonitrile, 2-nitro-4-trifluoromethyl, 2-cyano-5-trifluoromethyl nitrobenzene, 2-nitro-4-trifluoromethyl benzenecarbonitrile, 2-nitro-4-trifluoromethylbenzotrile, pubchem15190, acmc-1bf2x
PubChem CID 69902
IUPAC Name 2-nitro-4-(trifluoromethyl)benzonitrile
SMILES [O-][N+](=O)C1=CC(=CC=C1C#N)C(F)(F)F

Specifications

Melting Point 40°C to 44°C
Boiling Point 156°C to 158°C (18 mmHg)
Flash Point >110°C (230°F)
Quantity 25 g
UN Number UN3439
Beilstein 1882259
Formula Weight 216.12
Percent Purity 98%
Chemical Name or Material 2-Nitro-4-(trifluoromethyl)benzonitrile
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.