accessibility menu, dialog, popup

UserName

KKOSOOOSOOO

CAS RN 7790-62-7

IUPAC Name: dipotassium (sulfonatooxy)sulfonate
Synonyms: potassium pyrosulfate
Molecular Weight (g/mol): 254.31
Molecular Formula: K2O7S2
InChi Key: KAQHZJVQFBJKCK-UHFFFAOYSA-L
SMILES: [K+].[K+].[O-]S(=O)(=O)OS([O-])(=O)=O
Molecular Weight (g/mol) 
  • (6)
  • (2)
Quantity 
  • (1)
  • (1)
  • (2)
  • (1)
  • (3)
Physical Form 
  • (2)
  • (2)
  • (1)
  • (1)
Percent Purity 
  • (1)
  • (2)
Grade 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (4)
  • (6)
  • (2)
  • (1)
  • (1)
  • (2)
  • (1)
  • (3)
  • (2)
  • (2)
  • (1)
  • (1)
  • (1)
  • (2)
  • (2)